Alfa Aesar™ 3,5,5-Trimethylcyclohexane-1,2-dione, 99%

Manufacturer: Alfa Aesar™ B2262514

Quantity 25g

 View more versions of this product

Catalog No. AAB2262514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57696-89-6
    Molecular Formula C9H14O2
    Formula Weight 154.21
    MDL Number MFCD02181023
    Chemical Name or Material 3,5,5-Trimethylcyclohexane-1,2-dione

    Specifications

    Formula Weight 154.21
    TSCA TSCA
    Molecular Formula C9H14O2
    MDL Number MFCD02181023
    Flash Point >100°C (212°F)
    Melting Point 89° to 92°C
    Quantity 25g
    CAS 57696-89-6
    Chemical Name or Material 3,5,5-Trimethylcyclohexane-1,2-dione
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit