Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5,5-Trimethylcyclohexane-1,2-dione, 99%

Click to view available options
Quantity:
5g
25g
57696-89-6
C9H14O2
154.209
MFCD02181023
WEUZZYGESXCTCU-UHFFFAOYSA-N
3,5,5-trimethyl-1,2-cyclohexanedione,1,2-cyclohexanedione, 3,5,5-trimethyl,3,5,5-trimethyl-1.2-cyclohexanedione,acmc-1aooj,ksc274g5l,3,5,5-trimethyl-1,2 cyclohexanedione,3,5,5-trimethyl cyclohexane-1,2-dione,3,5,5-trimethylcyclohexane-1,2-dione 5g
93772
3,5,5-trimethylcyclohexane-1,2-dione
CC1CC(CC(=O)C1=O)(C)C
57696-89-6
C9H14O2
154.209
MFCD02181023
WEUZZYGESXCTCU-UHFFFAOYSA-N
3,5,5-trimethyl-1,2-cyclohexanedione,1,2-cyclohexanedione, 3,5,5-trimethyl,3,5,5-trimethyl-1.2-cyclohexanedione,acmc-1aooj,ksc274g5l,3,5,5-trimethyl-1,2 cyclohexanedione,3,5,5-trimethyl cyclohexane-1,2-dione,3,5,5-trimethylcyclohexane-1,2-dione 5g
93772
3,5,5-trimethylcyclohexane-1,2-dione
CC1CC(CC(=O)C1=O)(C)C
Flash Point >100°C (212°F)
Quantity 5g
Melting Point 89°C to 92°C
Formula Weight 154.21
Percent Purity 99%
Chemical Name or Material 3,5,5-Trimethylcyclohexane-1,2-dione