Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5,5-Trimethylhexyl Acetate 93.0+%, TCI America™

Click to view available options
Quantity:
25mL
500mL
58430-94-7
C11H22O2
186.295
MFCD00026336
DGKXDLCVQSQVBC-UHFFFAOYSA-N
Acetic Acid 3,5,5-Trimethylhexyl Ester
42745
3,5,5-trimethylhexyl acetate
CC(CCOC(=O)C)CC(C)(C)C
58430-94-7
C11H22O2
186.295
MFCD00026336
DGKXDLCVQSQVBC-UHFFFAOYSA-N
Acetic Acid 3,5,5-Trimethylhexyl Ester
42745
3,5,5-trimethylhexyl acetate
CC(CCOC(=O)C)CC(C)(C)C
Color Colorless
Boiling Point 209°C
Quantity 500mL
Formula Weight 186.30
Physical Form Clear Liquid at 20°C
Percent Purity ≥93.0% (GC)
Chemical Name or Material 3,5,5-Trimethylhexyl Acetate

EINECSNumber : (2)-0735

RTECSNumber : AK0840000

TSCA : Yes