Alfa Aesar™ 3,5,5-Trimethyloxazolidine-2,4-dione, 98%

Manufacturer: Alfa Aesar™ B2436203

Quantity 1g

 View more versions of this product

Catalog No. AAB2436203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 127-48-0
    Molecular Formula C6H9NO3
    Formula Weight 143.14
    MDL Number MFCD00047084
    Beilstein 121627
    Merck Index 14,9706
    Synonym Trimethadione
    Chemical Name or Material 3,5,5-Trimethyloxazolidine-2,4-dione

    Specifications

    Formula Weight 143.14
    Quantity 1g
    Molecular Formula C6H9NO3
    MDL Number MFCD00047084
    Beilstein 121627
    Merck Index 14,9706
    RTECS Number RQ2100000
    CAS 127-48-0
    Synonym Trimethadione
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H360
    May damage fertility or the unborn child.
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 78° to 80°C (5mmHg)
    Melting Point 45° to 48°C
    Chemical Name or Material 3,5,5-Trimethyloxazolidine-2,4-dione
    Assay 98%

    Description

    Solubility
    Soluble in water 50 g/L.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

    GHS H Statement
    H360
    May damage fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit