Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5,5-Trimethyloxazolidine-2,4-dione, 98%

Manufacturer:  Alfa Aesar™ B2436206

Quantity 5g

 View more versions of this product

Catalog No. AAB2436206

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Solubility
Soluble in water 50 g/L.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

127-48-0
143.14
121627
Trimethadione
C6H9NO3
MFCD00047084
14,9706
3,5,5-Trimethyloxazolidine-2,4-dione

Specifications

78° to 80°C (5mmHg)
5g
C6H9NO3
MFCD00047084
14,9706
3,5,5-Trimethyloxazolidine-2,4-dione
45° to 48°C
127-48-0
143.14
121627
Trimethadione
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : RQ2100000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit