Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Bis(methoxycarbonyl)benzeneboronic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
177735-55-6
C10H11BO6
238.002
MFCD11053854
WEJWFDLAZSVCJK-UHFFFAOYSA-N
3,5-bis methoxycarbonyl phenylboronic acid,3,5-bis methoxycarbonyl phenyl boronic acid,3,5-bis methoxycarbonyl benzeneboronic acid,3,5-bis methoxycarbonyl phenyl boronic acid,1,3-benzenedicarboxylic acid, 5-borono-, 1,3-dimethyl ester,3,5-di methoxycarbonyl phenylboronic acid,acmc-1c6yh,3,5-bis methoxycarbonyl phenyl-dihydroxyborane
19363104
[3,5-bis(methoxycarbonyl)phenyl]boronic acid
B(C1=CC(=CC(=C1)C(=O)OC)C(=O)OC)(O)O
177735-55-6
C10H11BO6
238.002
MFCD11053854
WEJWFDLAZSVCJK-UHFFFAOYSA-N
3,5-bis methoxycarbonyl phenylboronic acid,3,5-bis methoxycarbonyl phenyl boronic acid,3,5-bis methoxycarbonyl benzeneboronic acid,3,5-bis methoxycarbonyl phenyl boronic acid,1,3-benzenedicarboxylic acid, 5-borono-, 1,3-dimethyl ester,3,5-di methoxycarbonyl phenylboronic acid,acmc-1c6yh,3,5-bis methoxycarbonyl phenyl-dihydroxyborane
19363104
[3,5-bis(methoxycarbonyl)phenyl]boronic acid
B(C1=CC(=CC(=C1)C(=O)OC)C(=O)OC)(O)O
Applications
It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 250mg
Formula Weight 238
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3,5-Bis(methoxycarbonyl)benzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning