Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3',5'-Bis(trifluoromethyl)acetophenone, 98%

Manufacturer:  Alfa Aesar™ A1034103

Quantity 1g

 View more versions of this product

Catalog No. AAA1034103


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

30071-93-3
256.15
2384789
C10H6F6O
MFCD00009910
3',5'-Bis (trifluoromethyl)acetophenone

Specifications

30071-93-3
256.15
2384789
1g
3',5'-Bis (trifluoromethyl)acetophenone
C10H6F6O
MFCD00009910
85°C (8mmHg)
82°C (179°F)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit