Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Bis(trifluoromethyl)aniline, 98%

Click to view available options
Quantity:
10g
50g
250g
328-74-5
C8H5F6N
229.125
MFCD00000394
CDIDGWDGQGVCIB-UHFFFAOYSA-N
3,5-bis trifluoromethyl aniline,3,5-di trifluoromethyl aniline,benzenamine, 3,5-bis trifluoromethyl,3,5-bis trifluoromethyl benzenamine,3',5'-bis-trifluoromethyl aniline,3,5-bis-trifluoromethylaniline,alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine,3,5-bis-trifluoromethyl-phenylamine,aniline, 3,5-bis trifluoromethyl
9480
3,5-bis(trifluoromethyl)aniline
C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F
328-74-5
C8H5F6N
229.125
MFCD00000394
CDIDGWDGQGVCIB-UHFFFAOYSA-N
3,5-bis trifluoromethyl aniline,3,5-di trifluoromethyl aniline,benzenamine, 3,5-bis trifluoromethyl,3,5-bis trifluoromethyl benzenamine,3',5'-bis-trifluoromethyl aniline,3,5-bis-trifluoromethylaniline,alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine,3,5-bis-trifluoromethyl-phenylamine,aniline, 3,5-bis trifluoromethyl
9480
3,5-bis(trifluoromethyl)aniline
C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F
Applications
3,5-Bis(trifluoromethyl)aniline was used in the synthesis of N-[2-hydroxy-1-naphthylidene]-3, 5-bis(trifluoromethyl)aniline, Schiff′s base and 5,7-bis(trifluoromethyl)aniline. It was also used in the synthesis of N-1-phenylethyl-3,5-bis(trifluoromethyl)aniline via titanium-catalyzed hydroamination reaction and N,N′-bis[3,5-bis(tri-fluoromethyl)phenyl]thiourea, organocatalyst.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 250g
Flash Point 83°C (181°F)
Boiling Point 75°C to 78°C (10mmHg)
Odor Recognizable
UN Number UN2810
Beilstein 654318
Formula Weight 229.13
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)aniline

GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P260-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : ZE9800000

TSCA : TSCA