Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzamide, 97%

Click to view available options
Quantity:
1g
5g
25g
22227-26-5
C9H5F6NO
257.135
MFCD00015511
YNOPIKHMZIOWHS-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzamide,3,5-di trifluoromethyl benzamide,3,5-bis trifluoromethyl-benzamide,benzamide, 3,5-bis trifluoromethyl,acmc-1cglb,ynopikhmziowhs-uhfffaoysa,3,5-bis-trifluoromethyl-benzamide,3,5-bis-trifluoromethyl benzamide,benzamide,3,5-bis trifluoromethyl
519927
3,5-bis(trifluoromethyl)benzamide
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)N
22227-26-5
C9H5F6NO
257.135
MFCD00015511
YNOPIKHMZIOWHS-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzamide,3,5-di trifluoromethyl benzamide,3,5-bis trifluoromethyl-benzamide,benzamide, 3,5-bis trifluoromethyl,acmc-1cglb,ynopikhmziowhs-uhfffaoysa,3,5-bis-trifluoromethyl-benzamide,3,5-bis-trifluoromethyl benzamide,benzamide,3,5-bis trifluoromethyl
519927
3,5-bis(trifluoromethyl)benzamide
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)N
Quantity 5g
Melting Point 161°C to 163°C
Beilstein 4734142
Formula Weight 257.14
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning