Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%

Manufacturer: Alfa Aesar™ A1030014

Quantity 25g

 View more versions of this product

Catalog No. AAA1030014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39234-86-1
    Molecular Formula C8H3ClF6O2S
    Formula Weight 312.61
    MDL Number MFCD00014725
    UN Number UN3261
    Beilstein 3621218
    Chemical Name or Material 3,5-Bis (trifluoromethyl)benzenesulfonyl chloride

    Specifications

    Formula Weight 312.61
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Beilstein 3621218
    CAS 39234-86-1
    UN Number UN3261
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    MDL Number MFCD00014725
    Melting Point 34° to 38°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Molecular Formula C8H3ClF6O2S
    Quantity 25g
    Chemical Name or Material 3,5-Bis (trifluoromethyl)benzenesulfonyl chloride
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit