Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzyl chloride, 97%

Click to view available options
Quantity:
1g
5g
75462-59-8
C9H5ClF6
262.579
MFCD00009906
OINTXXMBRBLMHH-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzyl chloride,1-chloromethyl-3,5-bis trifluoromethyl benzene,3,5-bis-trifluoromethyl benzyl chloride,3,5-bis trifluoromethyl benzylchloride,3,5-di trifluoromethyl benzyl chloride,benzene, 1-chloromethyl-3,5-bis trifluoromethyl,3,5-bis trifluoromethyl-1-chloromethyl benzene,1,3-bis trifluoromethyl-5-chloromethyl benzene
123576
1-(chloromethyl)-3,5-bis(trifluoromethyl)benzene
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CCl
75462-59-8
C9H5ClF6
262.579
MFCD00009906
OINTXXMBRBLMHH-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzyl chloride,1-chloromethyl-3,5-bis trifluoromethyl benzene,3,5-bis-trifluoromethyl benzyl chloride,3,5-bis trifluoromethyl benzylchloride,3,5-di trifluoromethyl benzyl chloride,benzene, 1-chloromethyl-3,5-bis trifluoromethyl,3,5-bis trifluoromethyl-1-chloromethyl benzene,1,3-bis trifluoromethyl-5-chloromethyl benzene
123576
1-(chloromethyl)-3,5-bis(trifluoromethyl)benzene
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CCl
Applications
3,5-Bis(trifluoromethyl)benzyl chloride has been used in the preparation of 1-(azidomethyl)-3,5-bis-(trifluoromethyl)benzene. 1-(azidomethyl)-3,5-bis-(trifluoromethyl)benzene, in 94% yield by an efficient nucleophilic substitution reaction between 3,5-bis-(trifluoromethyl)benzyl chloride, 4, and sodium azide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Quantity 5g
Boiling Point 46°C (3mmHg)
Flash Point 71°C (161°F)
Melting Point 29°C to 33°C
UN Number UN3261
Beilstein 656502
Solubility Information Slightly soluble in water.
Formula Weight 262.58
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 278-216-1

TSCA : No

Recommended Storage : Ambient temperatures