Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzylamine, 97%

Click to view available options
Quantity:
1g
5g
25g
85068-29-7
C9H7F6N
243.152
MFCD00009909
DHVHORCFFOSRBP-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzylamine,3,5-bis trifluoromethyl phenyl methanamine,3,5-di trifluoromethyl benzylamine,3,5-ditrifluoromethylbenzylamine,mbt-bym,3,5-bis trifluoromethyl phenyl methylamine,benzenemethanamine, 3,5-bis trifluoromethyl,3,5-bis-trifluoromethyl-benzylamine
521099
[3,5-bis(trifluoromethyl)phenyl]methanamine
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN
85068-29-7
C9H7F6N
243.152
MFCD00009909
DHVHORCFFOSRBP-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzylamine,3,5-bis trifluoromethyl phenyl methanamine,3,5-di trifluoromethyl benzylamine,3,5-ditrifluoromethylbenzylamine,mbt-bym,3,5-bis trifluoromethyl phenyl methylamine,benzenemethanamine, 3,5-bis trifluoromethyl,3,5-bis-trifluoromethyl-benzylamine
521099
[3,5-bis(trifluoromethyl)phenyl]methanamine
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN
Flash Point 78°C (172°F)
Boiling Point 81°C to 83°C (13mmHg)
Quantity 5g
Melting Point 54°C to 60°C
UN Number UN3259
Beilstein 7209267
Formula Weight 243.15
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzylamine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.