Alfa Aesar™ 3,5-Bis(trifluoromethyl)styrene, 98%, stab.

Manufacturer: Alfa Aesar™ B2349903

Quantity 1g

 View more versions of this product

Catalog No. AAB2349903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 349-59-7
    Molecular Formula C10H6F6
    Formula Weight 240.15
    MDL Number MFCD00075538
    UN Number UN1993
    Chemical Name or Material 3,5-Bis(trifluoromethyl)styrene, stabilized

    Specifications

    Formula Weight 240.15
    Molecular Formula C10H6F6
    MDL Number MFCD00075538
    CAS 349-59-7
    UN Number UN1993
    Melting Point 7° to 8°C
    Boiling Point 59° to 61°C (20mmHg)
    Quantity 1g
    Flash Point 50°C (122°F)
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2° to 8°C
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 3,5-Bis(trifluoromethyl)styrene, stabilized
    Assay 98%

    Description

    Applications
    3,5-Bis(trifluoromethyl)styrene is used to produce other chemicals for example 1-(3, 5-bis-Trifluoromethyl-phenyl)-ethane-1, 2-diol.

    Solubility
    Sparingly soluble in water.

    Notes
    Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit