Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-tert-butyl-2-hydroxybenzaldehyde, 99%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
37942-07-7
C15H22O2
234.339
MFCD00191998
RRIQVLZDOZPJTH-UHFFFAOYSA-N
3,5-di-tert-butyl-2-hydroxybenzaldehyde,3,5-di-tert-butylsalicylaldehyde,3,5-bis 1,1-dimethylethyl-2-hydroxy-benzaldehyde,benzaldehyde, 3,5-bis 1,1-dimethylethyl-2-hydroxy,3,5-di-t-butylsalicylaldehyde,3,5-di-tert-butyl-2-hydroxy-benzaldehyde,benzaldehyde, 2-hydroxy, 3,5-di t-butyl,3,5-bis tert-butyl-2-hydroxybenzaldehyde,pubchem11247,acmc-1aete
688023
3,5-ditert-butyl-2-hydroxybenzaldehyde
CC(C)(C)C1=CC(=C(C(=C1)C=O)O)C(C)(C)C
37942-07-7
C15H22O2
234.339
MFCD00191998
RRIQVLZDOZPJTH-UHFFFAOYSA-N
3,5-di-tert-butyl-2-hydroxybenzaldehyde,3,5-di-tert-butylsalicylaldehyde,3,5-bis 1,1-dimethylethyl-2-hydroxy-benzaldehyde,benzaldehyde, 3,5-bis 1,1-dimethylethyl-2-hydroxy,3,5-di-t-butylsalicylaldehyde,3,5-di-tert-butyl-2-hydroxy-benzaldehyde,benzaldehyde, 2-hydroxy, 3,5-di t-butyl,3,5-bis tert-butyl-2-hydroxybenzaldehyde,pubchem11247,acmc-1aete
688023
3,5-ditert-butyl-2-hydroxybenzaldehyde
CC(C)(C)C1=CC(=C(C(=C1)C=O)O)C(C)(C)C
Applications
3,5-Di-tert-butyl-2-hydroxybenzaldehyde is used in the synthesis of Mn(III)-salen complex and its diamino precursor 5,6-diamino-5,6-dideoxy-1,2-O-isopropylidene-3-O-methyl-β-L-idofuranose, chiral Schiff base ligand for an enantioselective copper-catalyzed addition of phenyl acetylene to imines, chiral oxazolidine ligand for the enantioselective addition of diethyl zinc to aldehydes and tin Schiff base complexes with histidine analogues. It has antibacterial activity and is used in the preparation nickel complexes. It is structurally related to 3,5-di-t-butylcatechol (DTCAT) but is not as potent an activator of rat skeletal muscle ryanodine receptor Ca2+ channel (RyRC).

Solubility
Soluble in DMSO and methanol.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point 59°C to 61°C
Quantity 5g
Assay Percent Range 0.99
Beilstein 1877810
Formula Weight 234.34
Percent Purity 99%
Chemical Name or Material 3,5-Di-tert-butyl-2-hydroxybenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : Air Sensitive