Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-tert-butylsalicylic acid, 99%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
19715-19-6
C15H22O3
250.34
MFCD00059058
ZWQBZEFLFSFEOS-UHFFFAOYSA-N
3,5-di-tert-butyl-2-hydroxybenzoic acid,3,5-bis-tert-butylsalicylic acid,3,5-di-tert-butylsalicylic acid,benzoic acid, 3,5-bis 1,1-dimethylethyl hydroxy,benzoic acid, 3,5-bis 1,1-dimethylethyl-2-hydroxy,3,5-ditertbutyl salicylic acid,dibutylsalicylicacid,acmc-1c7qv,3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid,ksc466o9n
88208
3,5-ditert-butyl-2-hydroxybenzoic acid
CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C
19715-19-6
C15H22O3
250.34
MFCD00059058
ZWQBZEFLFSFEOS-UHFFFAOYSA-N
3,5-di-tert-butyl-2-hydroxybenzoic acid,3,5-bis-tert-butylsalicylic acid,3,5-di-tert-butylsalicylic acid,benzoic acid, 3,5-bis 1,1-dimethylethyl hydroxy,benzoic acid, 3,5-bis 1,1-dimethylethyl-2-hydroxy,3,5-ditertbutyl salicylic acid,dibutylsalicylicacid,acmc-1c7qv,3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid,ksc466o9n
88208
3,5-ditert-butyl-2-hydroxybenzoic acid
CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 158.0°C to 162.0°C
Flash Point 179°C
Color White to Beige
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ((CH3)3C)2C6H2(OH)COOH
Formula Weight 250.34
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3, 5-Di-tert-butylsalicylic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning