Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diaminobenzoic Acid Dihydrochloride 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
618-56-4
C7H8N2O2·2HCl
225.08
MFCD00012983
IGPLRBRBTUNCRT-UHFFFAOYSA-N
3,5-diaminobenzoic acid dihydrochloride,benzoic acid, 3,5-diamino-dihydrochloride,benzoic acid, 3,5-diamino-, hydrochloride 1:2,acmc-1b2s7,3,5-diaminobenzoicaciddihydrochloride,3,5-diamino benzoic acid dihydrochloride
15556789
3,5-diaminobenzoic acid;dihydrochloride
C1=C(C=C(C=C1N)N)C(=O)O.Cl.Cl
618-56-4
C7H8N2O2·2HCl
225.08
MFCD00012983
IGPLRBRBTUNCRT-UHFFFAOYSA-N
3,5-diaminobenzoic acid dihydrochloride,benzoic acid, 3,5-diamino-dihydrochloride,benzoic acid, 3,5-diamino-, hydrochloride 1:2,acmc-1b2s7,3,5-diaminobenzoicaciddihydrochloride,3,5-diamino benzoic acid dihydrochloride
15556789
3,5-diaminobenzoic acid;dihydrochloride
C1=C(C=C(C=C1N)N)C(=O)O.Cl.Cl
Melting Point >300.0°C
Quantity 25g
Color Beige to Gray
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (H2N)2C6H3CO2H·2HCl
Beilstein 14, 453
Solubility Information Solubility in water: 0.1 g/mL (20°C).
Formula Weight 225.08
Physical Form Fluffy Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material 3, 5-Diaminobenzoic acid dihydrochloride, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 210-556-8

RTECSNumber : DG6240000

TSCA : TSCA