Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3',5'-Dibenzyloxyacetophenone, 98%

Click to view available options
Quantity:
10g
50g
250g
28924-21-2
C22H20O3
332.399
MFCD00004777
KOJXGMJOTRYLBD-UHFFFAOYSA-N
3',5'-dibenzyloxyacetophenone,3,5-dibenzyloxyacetophenone,ethanone, 1-3,5-bis phenylmethoxy phenyl,3,5-bis benzyloxy acetophenone,1-3,5-bis phenylmethoxy phenyl ethanone,1-3,5-bis benzyloxy phenyl ethanone,3',5'-bis benzyloxy acetophenone,ethanone,1-3,5-bis phenylmethoxy phenyl
120098
1-[3,5-bis(phenylmethoxy)phenyl]ethanone
CC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3
28924-21-2
C22H20O3
332.399
MFCD00004777
KOJXGMJOTRYLBD-UHFFFAOYSA-N
3',5'-dibenzyloxyacetophenone,3,5-dibenzyloxyacetophenone,ethanone, 1-3,5-bis phenylmethoxy phenyl,3,5-bis benzyloxy acetophenone,1-3,5-bis phenylmethoxy phenyl ethanone,1-3,5-bis benzyloxy phenyl ethanone,3',5'-bis benzyloxy acetophenone,ethanone,1-3,5-bis phenylmethoxy phenyl
120098
1-[3,5-bis(phenylmethoxy)phenyl]ethanone
CC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3
Applications
3,5-Dibenzyloxyacetophenone is used in the synthesis of enantiomerically pure B-ring modified analogs of (-)-epicatechin gallate.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Melting Point 60°C to 62°C
Quantity 50g
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol
Formula Weight 332.4
Percent Purity 98%
Chemical Name or Material 3',5'-Dibenzyloxyacetophenone

EINECSNumber : 249-315-7

TSCA : Yes

Recommended Storage : Ambient temperatures