Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3',5'-Dibromo-2'-hydroxyacetophenone, 99%

Click to view available options
Quantity:
5g
25g
100g
22362-66-9
C8H6Br2O2
293.942
MFCD00191992
RCKHUOIKVIOMGZ-UHFFFAOYSA-N
3',5'-dibromo-2'-hydroxyacetophenone,1-3,5-dibromo-2-hydroxyphenyl ethanone,1-3,5-dibromo-2-hydroxyphenyl ethan-1-one,3´,5´-dibromo-2´-hydroxyacetophenone,1-acetyl-3,5-dibromo-2-hydroxybenzene,acmc-1cp7q,2-acetyl-4,6-dibromophenol,dibromooxochromenecarboxylicacid,3/',5/'-dibromo-2/'-hydroxyacetophenone,ethanone,1-3,5-dibromo-2-hydroxyphenyl
271762
1-(3,5-dibromo-2-hydroxyphenyl)ethanone
CC(=O)C1=CC(=CC(=C1O)Br)Br
22362-66-9
C8H6Br2O2
293.942
MFCD00191992
RCKHUOIKVIOMGZ-UHFFFAOYSA-N
3',5'-dibromo-2'-hydroxyacetophenone,1-3,5-dibromo-2-hydroxyphenyl ethanone,1-3,5-dibromo-2-hydroxyphenyl ethan-1-one,3´,5´-dibromo-2´-hydroxyacetophenone,1-acetyl-3,5-dibromo-2-hydroxybenzene,acmc-1cp7q,2-acetyl-4,6-dibromophenol,dibromooxochromenecarboxylicacid,3/',5/'-dibromo-2/'-hydroxyacetophenone,ethanone,1-3,5-dibromo-2-hydroxyphenyl
271762
1-(3,5-dibromo-2-hydroxyphenyl)ethanone
CC(=O)C1=CC(=CC(=C1O)Br)Br
Melting Point 108°C to 111°C
Quantity 100g
Formula Weight 293.95
Percent Purity 99%
Chemical Name or Material 3',5'-Dibromo-2'-hydroxyacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning