Learn More
3,5-dibromo-2-hydroxybenzaldehyde, 95%, Thermo Scientific™
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Linear Formula | Br2C6H2(OH)CHO |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 279.91 |
Percent Purity | 95% |
Chemical Name or Material | 3, 5-Dibromo-2-hydroxybenzaldehyde |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.