Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dibromo-4-methylbenzoic acid, 98+%

Click to view available options
Quantity:
1g
5g
67973-32-4
C8H6Br2O2
293.942
MFCD00017544
SJCBUASMFSRONS-UHFFFAOYSA-N
3,5-dibromo-p-toluic acid,benzoic acid,3,5-dibromo-4-methyl,benzoic acid, 3,5-dibromo-4-methyl,3,5-dibromo-4-methyl-benzoic acid,pubchem3897,timtec-bb sbb001202,3,5-dibromo-4-methylbenzoicacid,zerenex e/9072915
670475
3,5-dibromo-4-methylbenzoic acid
CC1=C(C=C(C=C1Br)C(=O)O)Br
67973-32-4
C8H6Br2O2
293.942
MFCD00017544
SJCBUASMFSRONS-UHFFFAOYSA-N
3,5-dibromo-p-toluic acid,benzoic acid,3,5-dibromo-4-methyl,benzoic acid, 3,5-dibromo-4-methyl,3,5-dibromo-4-methyl-benzoic acid,pubchem3897,timtec-bb sbb001202,3,5-dibromo-4-methylbenzoicacid,zerenex e/9072915
670475
3,5-dibromo-4-methylbenzoic acid
CC1=C(C=C(C=C1Br)C(=O)O)Br
Quantity 5g
Melting Point ~242°C
Beilstein 2577656
Formula Weight 293.94
Percent Purity ≥98%
Chemical Name or Material 3,5-Dibromo-4-methylbenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning