Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dibromobenzamide, 97%

Click to view available options
Quantity:
1g
5g
25g
175205-85-3
C7H5Br2NO
278.931
MFCD00178771
IOFGMNBDIDEBIQ-UHFFFAOYSA-N
3,5-dibromo-benzoic acid amide,acmc-20andi,3,5-dibromo-benzamide,maybridge1_007750,3,5-bis bromanyl benzamide,3,5-dibromobenzamide
2735937
3,5-dibromobenzamide
C1=C(C=C(C=C1Br)Br)C(=O)N
175205-85-3
C7H5Br2NO
278.931
MFCD00178771
IOFGMNBDIDEBIQ-UHFFFAOYSA-N
3,5-dibromo-benzoic acid amide,acmc-20andi,3,5-dibromo-benzamide,maybridge1_007750,3,5-bis bromanyl benzamide,3,5-dibromobenzamide
2735937
3,5-dibromobenzamide
C1=C(C=C(C=C1Br)Br)C(=O)N
Melting Point 180°C to 184°C
Quantity 1g
Formula Weight 278.93
Percent Purity 97%
Chemical Name or Material 3,5-Dibromobenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning