Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromobenzoic acid, 99%, Thermo Scientific™

Catalog Number AC352490050
Click to view available options
Quantity:
1 g
5 g
25 g
618-58-6
C7H4Br2O2
279.92
MFCD00051758
SFTFNJZWZHASAQ-UHFFFAOYSA-N
benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h
12063
3,5-dibromobenzoic acid
OC(=O)C1=CC(Br)=CC(Br)=C1
618-58-6
C7H4Br2O2
279.92
MFCD00051758
SFTFNJZWZHASAQ-UHFFFAOYSA-N
benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h
12063
3,5-dibromobenzoic acid
OC(=O)C1=CC(Br)=CC(Br)=C1
Melting Point 212.0°C to 214.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Linear Formula Br2C6H3CO2H
Quantity 5 g
Formula Weight 279.91
Percent Purity 99%
Chemical Name or Material 3,5-Dibromobenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit