Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-1,2,4-thiadiazole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2254-88-8
C2Cl2N2S
155.01
CEUHPOVLEQUFCC-UHFFFAOYSA-N
dichloro-1,2,4-thiadiazole,3,5-dichloro-1,2,4 thiadiazole,1,2,4-thiadiazole, 3,5-dichloro,1,2,4-thiadiazole,3,5-dichloro,3,5-dichloro-1,2,4 thiadiazol
2725055
3,5-dichloro-1,2,4-thiadiazole
C1(=NSC(=N1)Cl)Cl
2254-88-8
C2Cl2N2S
155.01
CEUHPOVLEQUFCC-UHFFFAOYSA-N
dichloro-1,2,4-thiadiazole,3,5-dichloro-1,2,4 thiadiazole,1,2,4-thiadiazole, 3,5-dichloro,1,2,4-thiadiazole,3,5-dichloro,3,5-dichloro-1,2,4 thiadiazol
2725055
3,5-dichloro-1,2,4-thiadiazole
C1(=NSC(=N1)Cl)Cl
Boiling Point 165.0°C to 167.0°C
Quantity 25g
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5580 to 1.5600
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 155.01
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3,5-Dichloro-1,2,4-thiadiazole
Show More Show Less

GHS Signal Word: Warning