Alfa Aesar™ 3,5-Dichloro-2,4,6-trifluoropyridine, 97+%

Manufacturer: Alfa Aesar™ A1020506

Quantity 5g

 View more versions of this product

Catalog No. AAA1020506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1737-93-5
    Molecular Formula C5Cl2F3N
    Formula Weight 201.96
    MDL Number MFCD00006226
    UN Number UN2811
    Beilstein 475874
    Chemical Name or Material 3,5-Dichloro-2,4,6-trifluoropyridine

    Specifications

    Formula Weight 201.96
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2811
    Molecular Formula C5Cl2F3N
    CAS 1737-93-5
    Beilstein 475874
    MDL Number MFCD00006226
    Boiling Point 159° to 160°C
    Melting Point 24° to 26°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H318-H315-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    TSCA TSCA
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 5g
    Chemical Name or Material 3,5-Dichloro-2,4,6-trifluoropyridine
    Assay ≥97%

    GHS H Statement
    H300-H310-H330-H318-H315-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit