Alfa Aesar™ 3,5-Dichloro-2-iodotoluene, 98%

Manufacturer: Alfa Aesar™ A1145206

Quantity 5g

 View more versions of this product

Catalog No. AAA1145206

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 175277-97-1
    Molecular Formula C7H5Cl2I
    Formula Weight 286.93
    MDL Number MFCD00052856
    Synonym 2,4-Dichloro-6-methyliodobenzene
    Chemical Name or Material 3,5-Dichloro-2-iodotoluene

    Specifications

    Formula Weight 286.93
    CAS 175277-97-1
    Molecular Formula C7H5Cl2I
    Melting Point 43° to 45°C
    Synonym 2,4-Dichloro-6-methyliodobenzene
    MDL Number MFCD00052856
    Quantity 5g
    Chemical Name or Material 3,5-Dichloro-2-iodotoluene
    Assay 98%

    Description

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit