Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dichloro-6-hydroxyaniline, ≥98%

Click to view available options
Quantity:
5g
25g
100g
527-62-8
C6H5Cl2NO
178.012
MFCD00035766
WASQBNCGNUTVNI-UHFFFAOYSA-N
2,4-dichloro-6-aminophenol,phenol, 2-amino-4,6-dichloro,unii-es86dyt9kh,2-amino-4,6-dichloro-phenol,ccris 7685,es86dyt9kh,6-amino-2,4-dichlorophenol,3,5-dichloro-2-hydroxyaniline,2,4-dichloro-6-amino phenol,acmc-209l1k
10699
2-amino-4,6-dichlorophenol
C1=C(C=C(C(=C1Cl)O)N)Cl
527-62-8
C6H5Cl2NO
178.012
MFCD00035766
WASQBNCGNUTVNI-UHFFFAOYSA-N
2,4-dichloro-6-aminophenol,phenol, 2-amino-4,6-dichloro,unii-es86dyt9kh,2-amino-4,6-dichloro-phenol,ccris 7685,es86dyt9kh,6-amino-2,4-dichlorophenol,3,5-dichloro-2-hydroxyaniline,2,4-dichloro-6-amino phenol,acmc-209l1k
10699
2-amino-4,6-dichlorophenol
C1=C(C=C(C(=C1Cl)O)N)Cl
Quantity 25g
Melting Point 89°C to 94°C
Color Yellow to Brown
UN Number UN2811
Beilstein 1102196
Formula Weight 178.01
Physical Form Solid
Percent Purity ≥98%
Chemical Name or Material 3,5-Dichloro-6-hydroxyaniline

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : SJ5774000