Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-N-methylbenzylamine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
90390-21-9
C8H9Cl2N
190.07
SSTJQZMMDIIKBR-UHFFFAOYSA-N
n-methyl-3,5-dichlorobenzylamine,3,5-dichlorophenyl methyl methyl amine,3,5-dichloro-n-methylbenzylamine,1-3,5-dichlorophenyl-n-methylmethanamine,benzenemethanamine, 3,5-dichloro-n-methyl,1-3,5-dichlorophenyl-n-methyl-methanamine,benzylamine, n-methyl-3,5-dichloro,90389-22-3 hydrochloride,3,5-dichloro-n-methyl-benzenemethanamine
146163
1-(3,5-dichlorophenyl)-N-methylmethanamine
CNCC1=CC(=CC(=C1)Cl)Cl
90390-21-9
C8H9Cl2N
190.07
SSTJQZMMDIIKBR-UHFFFAOYSA-N
n-methyl-3,5-dichlorobenzylamine,3,5-dichlorophenyl methyl methyl amine,3,5-dichloro-n-methylbenzylamine,1-3,5-dichlorophenyl-n-methylmethanamine,benzenemethanamine, 3,5-dichloro-n-methyl,1-3,5-dichlorophenyl-n-methyl-methanamine,benzylamine, n-methyl-3,5-dichloro,90389-22-3 hydrochloride,3,5-dichloro-n-methyl-benzenemethanamine
146163
1-(3,5-dichlorophenyl)-N-methylmethanamine
CNCC1=CC(=CC(=C1)Cl)Cl
Color Colorless to Yellow
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 190.07
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 3, 5-Dichloro-N-methylbenzylamine

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger