Alfa Aesar™ 3,5-Dichlorobenzamide, 97%

Manufacturer: Alfa Aesar™ B2195922

Quantity 100g

 View more versions of this product

Catalog No. AAB2195922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5980-23-4
    Molecular Formula C7H5Cl2NO
    Formula Weight 190.03
    MDL Number MFCD00057644
    Beilstein 2249768
    Chemical Name or Material 3,5-Dichlorobenzamide

    Specifications

    Formula Weight 190.03
    Molecular Formula C7H5Cl2NO
    MDL Number MFCD00057644
    CAS 5980-23-4
    Beilstein 2249768
    Melting Point 159° to 161°C
    Quantity 100g
    Chemical Name or Material 3,5-Dichlorobenzamide
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit