Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dichlorobenzamide, 97%

Click to view available options
Quantity:
5g
25g
100g
5980-23-4
C7H5Cl2NO
190.023
MFCD00057644
DELNZTRPJTUOIP-UHFFFAOYSA-N
benzamide,3,5-dichloro,benzamide, 3,5-dichloro,3,5-dichloro-benzamide,acmc-1b12b,3,5-bis chloranyl benzamide,3,5-dichloro benzamide
22289
3,5-dichlorobenzamide
C1=C(C=C(C=C1Cl)Cl)C(=O)N
5980-23-4
C7H5Cl2NO
190.023
MFCD00057644
DELNZTRPJTUOIP-UHFFFAOYSA-N
benzamide,3,5-dichloro,benzamide, 3,5-dichloro,3,5-dichloro-benzamide,acmc-1b12b,3,5-bis chloranyl benzamide,3,5-dichloro benzamide
22289
3,5-dichlorobenzamide
C1=C(C=C(C=C1Cl)Cl)C(=O)N
Quantity 25g
Melting Point 159°C to 161°C
Beilstein 2249768
Formula Weight 190.03
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzamide