Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dichlorobenzeneboronic acid, 98+%

Click to view available options
Quantity:
1g
5g
25g
67492-50-6
C6H5BCl2O2
190.814
MFCD00051935
DKYRKAIKWFHQHM-UHFFFAOYSA-N
3,5-dichlorophenyl boronic acid,3,5-dichlorobenzeneboronic acid,3,5-dichlorophenyl boranediol,3,5-dichlophenylboronic acid,3,5-dichloro phenylboronic acid,3,5-dichlorobenzene boronic acid,3,5-dichlorophenyl dihydroxyborane,boronic acid, b-3,5-dichlorophenyl,3,5-dichlorophenylboronicacid
2734331
(3,5-dichlorophenyl)boronic acid
B(C1=CC(=CC(=C1)Cl)Cl)(O)O
67492-50-6
C6H5BCl2O2
190.814
MFCD00051935
DKYRKAIKWFHQHM-UHFFFAOYSA-N
3,5-dichlorophenyl boronic acid,3,5-dichlorobenzeneboronic acid,3,5-dichlorophenyl boranediol,3,5-dichlophenylboronic acid,3,5-dichloro phenylboronic acid,3,5-dichlorobenzene boronic acid,3,5-dichlorophenyl dihydroxyborane,boronic acid, b-3,5-dichlorophenyl,3,5-dichlorophenylboronicacid
2734331
(3,5-dichlorophenyl)boronic acid
B(C1=CC(=CC(=C1)Cl)Cl)(O)O
Melting Point ~315°C (decomposition)
Quantity 1g
Beilstein 7581091
Formula Weight 190.82
Percent Purity ≥98%
Chemical Name or Material 3,5-Dichlorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning