Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3,5-Dichlorobenzeneboronic acid, 98+%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Melting Point | ~315°C (decomposition) |
Quantity | 25g |
Beilstein | 7581091 |
Formula Weight | 190.82 |
Percent Purity | ≥98% |
Chemical Name or Material | 3,5-Dichlorobenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures