Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dichlorobenzoyl chloride, 96%

Click to view available options
Quantity:
5g
25g
100g
2905-62-6
C7H3Cl3O
209.45
MFCD00009817
GGHLXLVPNZMBQR-UHFFFAOYSA-N
benzoyl chloride, 3,5-dichloro,3,5-dichlorobenzoylchloride,dichlorobenzoylchloride 3,5-,pubchem10805,dsstox_cid_7506,acmc-209h6l,dsstox_rid_78478,dsstox_gsid_27506,ksc204k6p,3,5-dichloro benzoyl chloride
76191
3,5-dichlorobenzoyl chloride
C1=C(C=C(C=C1Cl)Cl)C(=O)Cl
2905-62-6
C7H3Cl3O
209.45
MFCD00009817
GGHLXLVPNZMBQR-UHFFFAOYSA-N
benzoyl chloride, 3,5-dichloro,3,5-dichlorobenzoylchloride,dichlorobenzoylchloride 3,5-,pubchem10805,dsstox_cid_7506,acmc-209h6l,dsstox_rid_78478,dsstox_gsid_27506,ksc204k6p,3,5-dichloro benzoyl chloride
76191
3,5-dichlorobenzoyl chloride
C1=C(C=C(C=C1Cl)Cl)C(=O)Cl
Boiling Point 245°C
Quantity 5g
Melting Point 24°C to 28°C
Flash Point 137°C (278°F)
UN Number UN3261
Beilstein 1940681
Formula Weight 209.46
Percent Purity 96%
Chemical Name or Material 3,5-Dichlorobenzoyl chloride

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : DM6636766

TSCA : TSCA