Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorobenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
60211-57-6
C7H6Cl2O
177.03
MFCD00004634
VSNNLLQKDRCKCB-UHFFFAOYSA-N
3,5-dichlorobenzyl alcohol,3,5-dichlorophenyl methanol,benzenemethanol, 3,5-dichloro,3,5-dichlorobenzylalcohol,3,5-dichlorobenzylic alcohol,3,5-dichlorobenzenemethanol,3,5-dicholorbenzylalcohol,3,5-dichlorophenyl methan-1-ol,3, 5-dichlorobenzyl alcohol,pubchem4908
43236
(3,5-dichlorophenyl)methanol
C1=C(C=C(C=C1Cl)Cl)CO
60211-57-6
C7H6Cl2O
177.03
MFCD00004634
VSNNLLQKDRCKCB-UHFFFAOYSA-N
3,5-dichlorobenzyl alcohol,3,5-dichlorophenyl methanol,benzenemethanol, 3,5-dichloro,3,5-dichlorobenzylalcohol,3,5-dichlorobenzylic alcohol,3,5-dichlorobenzenemethanol,3,5-dicholorbenzylalcohol,3,5-dichlorophenyl methan-1-ol,3, 5-dichlorobenzyl alcohol,pubchem4908
43236
(3,5-dichlorophenyl)methanol
C1=C(C=C(C=C1Cl)Cl)CO
Infrared Spectrum Authentic
Quantity 5g
Melting Point 76°C to 80°C
Packaging Glass bottle
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula Cl2C6H3CH2OH
Formula Weight 177.03
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3, 5-Dichlorobenzyl alcohol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 262-109-1

TSCA : TSCA