Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Dichlorobiphenyl-4-sulfonyl chloride, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
885267-95-8
C12H7Cl3O2S
321.61
KBMKQHVXGMOKKI-UHFFFAOYSA-N
4-3,5-dichlorophenyl benzenesulfonyl chloride,3',5'-dichloro-1,1'-biphenyl-4-sulfonyl chloride,4-3,5-dichlorophenyl benzenesulphonyl chloride,3',5'-dichlorobiphenyl-4-sulfonyl chloride,4-3,5-dichlorophenyl phenyl sulfonyl chloride
3771398
4-(3,5-dichlorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC(=CC(=C2)Cl)Cl)S(=O)(=O)Cl
885267-95-8
C12H7Cl3O2S
321.61
KBMKQHVXGMOKKI-UHFFFAOYSA-N
4-3,5-dichlorophenyl benzenesulfonyl chloride,3',5'-dichloro-1,1'-biphenyl-4-sulfonyl chloride,4-3,5-dichlorophenyl benzenesulphonyl chloride,3',5'-dichlorobiphenyl-4-sulfonyl chloride,4-3,5-dichlorophenyl phenyl sulfonyl chloride
3771398
4-(3,5-dichlorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC(=CC(=C2)Cl)Cl)S(=O)(=O)Cl
Melting Point 102°C to 106°C
Quantity 5g
Assay Percent Range 96% min. diethylsulfonamide derivative (HPLC)
Formula Weight 321.61
Percent Purity 97%
Chemical Name or Material 3', 5'-Dichlorobiphenyl-4-sulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS Signal Word: Danger