Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3,5-Dichlorophenyl)methanesulfonyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
163295-70-3
C7H5Cl3O2S
259.54
MFCD01631928
GZMJRPMBVICCFL-UHFFFAOYSA-N
3,5-dichlorophenyl methanesulfonyl chloride,3,5-dichloro-phenyl-methanesulfonyl chloride,acmc-1c80o,3,5-dichloro-,a-toluenesufonyl chloride,3,5-dichlorophenyl methyl chlorosulfone,3,5-dichlorophenyl methanesulfonylchloride,3,5-dichloro-alpha-toluenesulfonyl chloride,3,5-dichlorophenyl methane-sulfonyl chloride,benzenemethanesulfonylchloride, 3,5-dichloro,3,5-dichlorophenyl methyl sulfonyl chloride
4993270
(3,5-dichlorophenyl)methanesulfonyl chloride
C1=C(C=C(C=C1Cl)Cl)CS(=O)(=O)Cl
163295-70-3
C7H5Cl3O2S
259.54
MFCD01631928
GZMJRPMBVICCFL-UHFFFAOYSA-N
3,5-dichlorophenyl methanesulfonyl chloride,3,5-dichloro-phenyl-methanesulfonyl chloride,acmc-1c80o,3,5-dichloro-,a-toluenesufonyl chloride,3,5-dichlorophenyl methyl chlorosulfone,3,5-dichlorophenyl methanesulfonylchloride,3,5-dichloro-alpha-toluenesulfonyl chloride,3,5-dichlorophenyl methane-sulfonyl chloride,benzenemethanesulfonylchloride, 3,5-dichloro,3,5-dichlorophenyl methyl sulfonyl chloride
4993270
(3,5-dichlorophenyl)methanesulfonyl chloride
C1=C(C=C(C=C1Cl)Cl)CS(=O)(=O)Cl
Infrared Spectrum Authentic
Melting Point 108.6°C to 110.6°C
Quantity 1g
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. as diethylsulfonamide derivative (HPLC)
Formula Weight 259.54
Physical Form Powder or Solid
Percent Purity 97%
Chemical Name or Material (3,5-Dichlorophenyl)methanesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger