Alfa Aesar™ 3,5-Dichlorosalicylic acid, 99%

Manufacturer: Alfa Aesar™ B2364114

Quantity 25g

 View more versions of this product

Catalog No. AAB2364114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 320-72-9
    Molecular Formula C7H4Cl2O3
    Formula Weight 207.01
    MDL Number MFCD00002442
    Synonym 3,5-Dichloro-2-hydroxybenzoic acid
    Chemical Name or Material 3,5-Dichlorosalicylic acid

    Specifications

    Formula Weight 207.01
    TSCA TSCA
    Melting Point 222° to 225°C
    Molecular Formula C7H4Cl2O3
    MDL Number MFCD00002442
    CAS 320-72-9
    Synonym 3,5-Dichloro-2-hydroxybenzoic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number VO2450000
    Quantity 25g
    Chemical Name or Material 3,5-Dichlorosalicylic acid
    Assay 99%

    Description

    Applications
    3,5-Dichlorosalicylic acid is a bio-active drug and is a potential inhibitor of human 20α-hydroxysteroid dehydrogenase. It also finds its application as medicine and dyes intermediates.

    Solubility
    Solubility in methanol (almost transparency).

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit