Alfa Aesar™ 3,5-Difluoro-2-methoxyaniline, 97%

Manufacturer: Alfa Aesar™ H3181603

Quantity 1g

 View more versions of this product

Catalog No. AAH3181603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41860-67-7
    Molecular Formula C7H7F2NO
    Formula Weight 159.14
    MDL Number MFCD09832386
    UN Number UN2810
    Synonym 3,5-Difluoro-o-anisidine
    Chemical Name or Material 3,5-Difluoro-2-methoxyaniline

    Specifications

    Formula Weight 159.14
    MDL Number MFCD09832386
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Molecular Formula C7H7F2NO
    CAS 41860-67-7
    UN Number UN2810
    Synonym 3,5-Difluoro-o-anisidine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Quantity 1g
    Chemical Name or Material 3,5-Difluoro-2-methoxyaniline
    Assay 97%

    GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit