Alfa Aesar™ 3,5-Difluoro-2-methylbenzoic acid, 97%

Manufacturer: Alfa Aesar™ H3161603

Quantity 1g

 View more versions of this product

Catalog No. AAH3161603

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C8H6F2O2
    Formula Weight 172.13
    MDL Number MFCD09832334
    Chemical Name or Material 3,5-Difluoro-2-methylbenzoic acid

    Specifications

    Formula Weight 172.13
    Molecular Formula C8H6F2O2
    MDL Number MFCD09832334
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 133° to 136°C
    Quantity 1g
    Chemical Name or Material 3,5-Difluoro-2-methylbenzoic acid
    Assay 97%

    Description

    Applications
    The mineralization of acidic aqueous solutions with 230 and 115 ppm 3,6-dichloro-2-methoxybenzoic acid (dicamba) in 0.05 M Na2SO4 of pH 3.0 has been studied by electro-Fenton and photoelectro-Fenton using a Pt anode and an O2-diffusion cathode, where oxidizing hydroxyl radicals are produced from Fenton ft.s reaction between added Fe2+ and H2O2 generated by the cathode. The fast mineralization of such complexes by UV light explains the highest oxidative ability of photoelectro-Fenton.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit