Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluoro-2-methylbenzonitrile, 97%, Thermo Scientific™

Catalog Number AAH34195MD
Click to view available options
Quantity:
250 mg
1 g
1003708-74-4
C8H5F2N
153.132
MFCD09996934
JHMDKULXRYFRBP-UHFFFAOYSA-N
3,5-difluoro-2-methyl benzonitrile, pubchem8342
44887034
3,5-difluoro-2-methylbenzonitrile
CC1=C(C=C(C=C1C#N)F)F
1003708-74-4
C8H5F2N
153.132
MFCD09996934
JHMDKULXRYFRBP-UHFFFAOYSA-N
3,5-difluoro-2-methyl benzonitrile, pubchem8342
44887034
3,5-difluoro-2-methylbenzonitrile
CC1=C(C=C(C=C1C#N)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 55°C to 57°C
Color White
Quantity 250 mg
UN Number UN3439
Formula Weight 153.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluoro-2-methylbenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit