Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Difluoro-4-methoxybenzoic acid, 97%

Click to view available options
Quantity:
250mg
1g
319-60-8
C8H6F2O3
188.13
MFCD01090994
YOWQTTYAQQPSHM-UHFFFAOYSA-N
3,5-difluoro-4-methoxybenzoicacid,3,5-difluoro-4-methoxy benzoic acid,4-carboxy-2,6-difluoroanisole,benzoic acid, 3,5-difluoro-4-methoxy,3,5-difluoro-4-methoxy-benzoic acid,acmc-20aic4,3,5difluoro-4-methoxy benzoic acid,3,5 difluoro-4-methoxy benzoic acid,benzoic acid,3,5-difluoro-4-methoxy,3,5-bis fluoranyl-4-methoxy-benzoic acid
2736987
3,5-difluoro-4-methoxybenzoic acid
COC1=C(C=C(C=C1F)C(=O)O)F
319-60-8
C8H6F2O3
188.13
MFCD01090994
YOWQTTYAQQPSHM-UHFFFAOYSA-N
3,5-difluoro-4-methoxybenzoicacid,3,5-difluoro-4-methoxy benzoic acid,4-carboxy-2,6-difluoroanisole,benzoic acid, 3,5-difluoro-4-methoxy,3,5-difluoro-4-methoxy-benzoic acid,acmc-20aic4,3,5difluoro-4-methoxy benzoic acid,3,5 difluoro-4-methoxy benzoic acid,benzoic acid,3,5-difluoro-4-methoxy,3,5-bis fluoranyl-4-methoxy-benzoic acid
2736987
3,5-difluoro-4-methoxybenzoic acid
COC1=C(C=C(C=C1F)C(=O)O)F
Quantity 250mg
Melting Point 164°C to 166°C
Formula Weight 188.13
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-4-methoxybenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures