Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Difluoro-4-methoxybenzyl bromide, 97%

Click to view available options
Quantity:
1g
5g
706786-42-7
C8H7BrF2O
237.044
MFCD04115916
IQTBMXJVMUSGSL-UHFFFAOYSA-N
3,5-difluoro-4-methoxybenzyl bromide,5-bromomethyl-1,3-difluoro-2-methoxybenzene,4-bromomethyl-2,6-difluoroanisole,benzene, 5-bromomethyl-1,3-difluoro-2-methoxy
17750664
5-(bromomethyl)-1,3-difluoro-2-methoxybenzene
COC1=C(C=C(C=C1F)CBr)F
706786-42-7
C8H7BrF2O
237.044
MFCD04115916
IQTBMXJVMUSGSL-UHFFFAOYSA-N
3,5-difluoro-4-methoxybenzyl bromide,5-bromomethyl-1,3-difluoro-2-methoxybenzene,4-bromomethyl-2,6-difluoroanisole,benzene, 5-bromomethyl-1,3-difluoro-2-methoxy
17750664
5-(bromomethyl)-1,3-difluoro-2-methoxybenzene
COC1=C(C=C(C=C1F)CBr)F
Applications
3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
Quantity 1g
Melting Point 39°C to 42°C
UN Number UN3261
Solubility Information Insoluble in water.
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-4-methoxybenzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures