Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Difluorobenzamide, 97%

Click to view available options
Quantity:
1g
5g
132980-99-5
C7H5F2NO
157.12
MFCD00061138
CGOIBYYWOVRGGV-UHFFFAOYSA-N
benzamide, 3,5-difluoro-9ci,benzamide,3,5-difluoro,acmc-20a3q0,3,5-bis fluoranyl benzamide,3,5-difluorobenzamide
2736910
3,5-difluorobenzamide
C1=C(C=C(C=C1F)F)C(=O)N
132980-99-5
C7H5F2NO
157.12
MFCD00061138
CGOIBYYWOVRGGV-UHFFFAOYSA-N
benzamide, 3,5-difluoro-9ci,benzamide,3,5-difluoro,acmc-20a3q0,3,5-bis fluoranyl benzamide,3,5-difluorobenzamide
2736910
3,5-difluorobenzamide
C1=C(C=C(C=C1F)F)C(=O)N
Melting Point 155°C to 157°C
Quantity 1g
Beilstein 4177620
Formula Weight 157.12
Percent Purity 97%
Chemical Name or Material 3,5-Difluorobenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures