Alfa Aesar™ 3,5-Difluorobenzhydrol, 95%

Manufacturer: Alfa Aesar™ H3184803

Quantity 1g

 View more versions of this product

Catalog No. AAH3184803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 153877-56-6
    Molecular Formula C13H10F2O
    Formula Weight 220.22
    MDL Number MFCD06201256
    Chemical Name or Material 3,5-Difluorobenzhydrol

    Specifications

    Formula Weight 220.22
    Molecular Formula C13H10F2O
    MDL Number MFCD06201256
    CAS 153877-56-6
    Quantity 1g
    Chemical Name or Material 3,5-Difluorobenzhydrol
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit