Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Difluorobenzoic acid, 97%

Click to view available options
Quantity:
5g
25g
455-40-3
C7H4F2O2
158.104
MFCD00010323
GONAVIHGXFBTOZ-UHFFFAOYSA-N
3,5-difluorobenzoicacid,benzoic acid, 3,5-difluoro,pubchem2173,3,5 difluorobenzoic acid,3,5-difluoro benzoicacid,acmc-209k3a,3,5 difluoro benzoic acid,3,5-di-fluorobenzoic acid,3,5-difluoro-benzoic acid
101265
3,5-difluorobenzoic acid
C1=C(C=C(C=C1F)F)C(=O)O
455-40-3
C7H4F2O2
158.104
MFCD00010323
GONAVIHGXFBTOZ-UHFFFAOYSA-N
3,5-difluorobenzoicacid,benzoic acid, 3,5-difluoro,pubchem2173,3,5 difluorobenzoic acid,3,5-difluoro benzoicacid,acmc-209k3a,3,5 difluoro benzoic acid,3,5-di-fluorobenzoic acid,3,5-difluoro-benzoic acid
101265
3,5-difluorobenzoic acid
C1=C(C=C(C=C1F)F)C(=O)O
Quantity 25g
Melting Point 119°C to 123°C
Odor Odorless
Beilstein 1940680
Formula Weight 158.11
Percent Purity 97%
Chemical Name or Material 3,5-Difluorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes