Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Difluorobenzyl alcohol, 97%

Click to view available options
Quantity:
1g
5g
25g
79538-20-8
C7H6F2O
144.121
MFCD00010313
LOGIHEKXJKHXEC-UHFFFAOYSA-N
3,5-difluorobenzyl alcohol,3,5-difluorophenyl methanol,benzenemethanol, 3,5-difluoro,3,5-difluorophenyl methan-1-ol,3,5-difluorobenzylalcohol,3, 5-difluorobenzyl alcohol,pubchem4918,acmc-1baxh,ksc493g5j,3,5-difluoro-phenyl methanol
522721
(3,5-difluorophenyl)methanol
C1=C(C=C(C=C1F)F)CO
79538-20-8
C7H6F2O
144.121
MFCD00010313
LOGIHEKXJKHXEC-UHFFFAOYSA-N
3,5-difluorobenzyl alcohol,3,5-difluorophenyl methanol,benzenemethanol, 3,5-difluoro,3,5-difluorophenyl methan-1-ol,3,5-difluorobenzylalcohol,3, 5-difluorobenzyl alcohol,pubchem4918,acmc-1baxh,ksc493g5j,3,5-difluoro-phenyl methanol
522721
(3,5-difluorophenyl)methanol
C1=C(C=C(C=C1F)F)CO
Applications
3,5-Difluorobenzyl alcohol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Refractive Index 1.487
Density 1.276
Flash Point 94°C (201°F)
Quantity 1g
Boiling Point 98°C to 99°C (9mmHg)
Beilstein 5499788
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 3,5-Difluorobenzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures