Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluoropyridine, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
71902-33-5
C5H3F2N
115.08
WRXAZPPGFLETFR-UHFFFAOYSA-N
pyridine, 3,5-difluoro,3,5-difluoro-pyridine,abbypharma ap-14-5380,3,5-difluoropyridine;,pubchem10556,acmc-209okl,3,5-difluoro-pyridine;,ksc377a3n,3,5-difluoropyridine,3,5-difluoro pyridine
642820
3,5-difluoropyridine
C1=C(C=NC=C1F)F
71902-33-5
C5H3F2N
115.08
WRXAZPPGFLETFR-UHFFFAOYSA-N
pyridine, 3,5-difluoro,3,5-difluoro-pyridine,abbypharma ap-14-5380,3,5-difluoropyridine;,pubchem10556,acmc-209okl,3,5-difluoro-pyridine;,ksc377a3n,3,5-difluoropyridine,3,5-difluoro pyridine
642820
3,5-difluoropyridine
C1=C(C=NC=C1F)F
Packaging Glass Bottle
Specific Gravity 1.256
Quantity 1g
Flash Point 7°C
Infrared Spectrum Authentic
Density 1.2560g/mL
Boiling Point 92.0°C to 94.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 115.08
Percent Purity 97%
Chemical Name or Material 3,5-Difluoropyridine
Show More Show Less

GHS H Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Danger