Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Difluorothiobenzamide, 97%

Manufacturer:  Alfa Aesar™ H3171906

Quantity 5g

 View more versions of this product

Catalog No. AAH3171906


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

874791-10-3
173.18
UN2811
C7H5F2NS
MFCD04036082
3,5-Difluorothiobenzamide

Specifications

135° to 139°C
874791-10-3
173.18
UN2811
97%
5g
C7H5F2NS
MFCD04036082
3,5-Difluorothiobenzamide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit