Alfa Aesar™ 3,5-Dihydroxybenzoic acid, 98%

Manufacturer: Alfa Aesar™ A116640E

Quantity 2500g

 View more versions of this product

Catalog No. AAA116640E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99-10-5
    Molecular Formula C7H6O4
    Formula Weight 154.12
    MDL Number MFCD00002512
    Beilstein 2207864
    Synonym alpha-Resorcylic acid
    Chemical Name or Material 3,5-Dihydroxybenzoic acid

    Specifications

    Formula Weight 154.12
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym alpha-Resorcylic acid
    Quantity 2500g
    CAS 99-10-5
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Molecular Formula C7H6O4
    Beilstein 2207864
    Melting Point ≈237°C (dec.)
    MDL Number MFCD00002512
    RTECS Number VH3708000
    Chemical Name or Material 3,5-Dihydroxybenzoic acid
    Assay 98%

    Description

    Applications
    3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.

    Solubility
    Soluble in 50 mg/mL, 95% Ethanol.

    Notes
    Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit