Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dihydroxybenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
29654-55-5
C7H8O3
140.14
MFCD00016867
NGYYFWGABVVEPL-UHFFFAOYSA-N
3,5-dihydroxybenzyl alcohol,5-hydroxymethyl-1,3-benzenediol,5-hydroxymethyl benzene-1,3-diol,5-hydroxymethyl resorcinol,benzyl alcohol, 3,5-dihydroxy,1,3-benzenediol, 5-hydroxymethyl,3,5-dihydroxybenzylalcohol,3,5-dihydroxy benzyl alcohol,pubchem7418,acmc-1cjog
34661
5-(hydroxymethyl)benzene-1,3-diol
C1=C(C=C(C=C1O)O)CO
29654-55-5
C7H8O3
140.14
MFCD00016867
NGYYFWGABVVEPL-UHFFFAOYSA-N
3,5-dihydroxybenzyl alcohol,5-hydroxymethyl-1,3-benzenediol,5-hydroxymethyl benzene-1,3-diol,5-hydroxymethyl resorcinol,benzyl alcohol, 3,5-dihydroxy,1,3-benzenediol, 5-hydroxymethyl,3,5-dihydroxybenzylalcohol,3,5-dihydroxy benzyl alcohol,pubchem7418,acmc-1cjog
34661
5-(hydroxymethyl)benzene-1,3-diol
C1=C(C=C(C=C1O)O)CO
Melting Point 181.0°C to 186.0°C
Color Beige-Pink to White or Gray
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula (HO)2C6H3CH2OH
Formula Weight 140.14
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3, 5-Dihydroxybenzyl alcohol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 249-751-8