Alfa Aesar™ 3,5-Dihydroxytoluene monohydrate, 97%

Manufacturer: Alfa Aesar™ A1044022

Quantity 100g

 View more versions of this product

Catalog No. AAA1044022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6153-39-5
    Molecular Formula C7H8O2·H2O
    Formula Weight 142.16 (124.14 Anhydrous)
    MDL Number MFCD00149092
    Beilstein 5103474
    Merck Index 14,6864
    Synonym 5-Methylresorcinol monohydrate; Orcinol monohydrate
    Chemical Name or Material 3,5-Dihydroxytoluene monohydrate

    Specifications

    Formula Weight 142.16 (124.14 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 6153-39-5
    Merck Index 14,6864
    Synonym 5-Methylresorcinol monohydrate; Orcinol monohydrate
    TSCA TSCA
    MDL Number MFCD00149092
    Beilstein 5103474
    Health Hazard 1 Warning
    Melting Point 56° to 60°C
    Molecular Formula C7H8O2·H2O
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number VH2100000
    Boiling Point 147°C (5mmHg)
    Quantity 100g
    Chemical Name or Material 3,5-Dihydroxytoluene monohydrate
    Assay 97%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit